Brady® i5100 IP 1 Core R6000 Series Printer Ribbon
Supplier: Brady Worldwide
Certificates
About this item
IP-Enabled ribbons come spooled on a specially designed core that includes a Smart Cell RFID tag that communicates with the Brady IP® industrial label printer.
- Halogen-free
- Heat-resistant
- Scratch-resistant
- Solvent-resistant
One-color ribbon for printing a single color onto a label. R6000 Series Halogen-free resin formulation offers excellent solvent, scratch, and heat resistance when printed on the appropriate label material. Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra aggressive polyester and B-423 Permanent polyester from Brady®. The Brady IP® industrial label printer status monitor shows the printer and material information on the user's PC.
These ribbons are designed for use with Brady IP® Series Label Printer and BradyPrinter i5100 Industrial Label Printer.
Frequently Bought Together