Digital Cel-Gro Tissue Culture Rotator, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
The Digital Cel-Gro Tissue Culture Rotator has a brushless no-maintenance motor that provides a gentle rotating motion for optimum results.
- Digital display of speed and time
- Speed range: 5 to 70 rpm
- Speed accuracy: ±1 rpm
- Adjustable rocking angle: 95° to 180°
- Loading capacity: 5 kg
- Timer range: 0 to 9999 minutes
- Optional multiple size drums offering different capacity of tube rotation
The Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotator precisely mixes test tube contents. Achieve optimum culture results for tissue and virus growth, hormone production, and sample testing of viruses and cytotoxicity assays.
Certifications: cCSAus.
Ordering information: Order minimum 1 drum together with the rotator. 2 years of warranty.
Specifications
- Catalog No:
- 77041-358
- Speed accuracy:
- ±1 rpm
- Platform Material:
- Stainless steel
- Display:
- Digital
- Control:
- Digital
Specifications
Frequently Bought Together