ESC Swabs, Liofilchem, Inc.
Supplier: LIOFILCHEM, INC.
Certificates
About this item
ESC (Easy Surface Checking) for verification and validation of cleaning procedures; cost effective and rapid method for microbial environmental monitoring.
- Prefilled swab device
- Rayon bud
- Wide range of transport media and volumes
- Leak-proof closure
- Transport and storage at room temperature
ESC (Easy Surface Checking) swab range includes a new generation of sampling devices for microbiological examination of surfaces, especially recommended for food and pharmaceutical industries.
Certifications: Conform to international standards (ISO 18593, ISO 17604)
Frequently Bought Together