Clean Tests, Liofilchem Inc.
Supplier: LIOFILCHEM, INC.
Certificates
About this item
Swab for detecting protein, fat and sugar residues for the hygiene control of surfaces.
- Results within 10 minutes
- Easy color change interpretation
- Minimal training required
Clean test is a swab for the detection of protein, fat and sugar residues suitable for the check of work surfaces cleaning level.
Frequently Bought Together