NEBNext® Cell Lysis Buffer Module, New England Biolabs
Catalog # 76326-030
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:NEBNext® Cell lysis buffer module
- Cat. No.:76326-030
- No. of reactions:96
Specifications
About this item
This module contains the enzymes and buffers required to lyse isolated cultured and primary cells to extract RNA. The module is optimized for use with the NEBNext Single Cell/Low Input RNA Library Prep Kit for Illumina or NEBNext Single Cell/Low Input cDNA Synthesis and Amplification Module, and can be used to supplement the NEBNext Cell Lysis Buffer and Murine RNase Inhibitor components supplied in these products.
- Includes NEBNext Cell Lysis Buffer
- Includes Murine RNase Inhibitor