Order Entry
United States
ContactUsLinkComponent
NEBuffer™ Set (r1.1, r2.1, r3.1 and rCutSmart™), New England Biolabs
NEBuffer™ Set (r1.1, r2.1, r3.1 and rCutSmart™), New England Biolabs
Catalog # 76486-070
CAS Number:  
undefined
NEBuffer™ Set (r1.1, r2.1, r3.1 and rCutSmart™), New England Biolabs
Catalog # 76486-070
Supplier Number:  B7030S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    10X
  • Description:
    NEBuffer™ Set (r1.1, r2.1, r3.1 and rCutSmart™)
  • Size:
    1.25 ml each
  • Cat. No.:
    76486-070

Specifications

About this item

New England Biolabs provides 10X NEBuffer with each restriction endonuclease to assure optimal (100%) activity. Most of our enzymes are supplied with one of four standard NEBuffers. This set contains NEBuffers that contain recombinant Albumin.

  • Contains recombinant albumin
  • Set contains 1.25 ml vial of each buffer