Phenol Red Broth with Galactose
Prepared media (Phenol red broth with galactose), 10 ml, 20/pack, 16×125 mm, Tube with Durham tube
Catalog # 76375-808
Supplier: Hardy Diagnostics
: Y306
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
For the differentiation of microorganisms by carbohydrate fermentation. Contains a durham tube for gas detection.
Specifications
Specifications
- Pack type:Tube with Durham tube
- Description:Prepared media (Phenol red broth with galactose)
- Size:20/pack
- Tube size:16×125 mm
- Volume:10 ml



