Order Entry
United States
ContactUsLinkComponent
Mission Complete Linearity Control, Diamond Diagnostics
Mission Complete Linearity Control, Diamond Diagnostics
Catalog # 103881-370
CAS Number:  
undefined
Mission Complete Linearity Control, Diamond Diagnostics
Catalog # 103881-370
Supplier Number:  DD-92900
CAS Number:  

Specifications

  • Description:
    Mission complete linearity control
  • Cat. No.:
    103881-370

Specifications

About this item

Mission complete linearity, metabolite and cooximetry controls.

  • Contains 4 vials of each of 5 levels at 1.8 ml
  • For use on all blood, gas and electrolyte systems
  • Assayed for pH, pCO₂, pO₂, Na⁺, K⁺, Ca⁺⁺, Cl⁻, Li⁺ glucose and lactate

Compatible with Medica Easylyte series, Diamond ProLyte®, Smartlyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, IL ILyte and Menarini Spotlyte, Alfa Wassermann Starlyte III, Starlyte V, Roche 9110, 9120, 9130, 9140, 9180, 9181, Radiometer ABL 5, 50, 500, 510, 520, 555, EML 100, 105, Siemens Rapidlab 248, 348, 840, 845, 850, 855, 860, 865, Roche AVL Compact 1-2-3.