ContactUsLinkComponent
Todd Hewitt Broth
Todd Hewitt Broth
Catalog # 76377-546
Supplier:  Hardy Diagnostics
CAS Number:  
undefined
Todd Hewitt Broth
Catalog # 76377-546
Supplier:  Hardy Diagnostics
Supplier Number:  K112
CAS Number:  
Specifications
  • Pack type:
    Glass tube
  • Description:
    Prepared media
  • Size:
    20/pack
  • Minimum Order Quantity:
    5 packs of 20
  • Cat. No.:
    76377-546
  • Tube size:
    13×100 mm
Specifications
About this item
For the cultivation of Streptococcus spp. for serological typing.