Order Entry
United States
ContactUsLinkComponent
Lithium acetate 1 mol/l in aqueous solution
Lithium acetate 1 mol/l in aqueous solution
Catalog # 82023-112
Supplier:  G-Biosciences
CAS Number:  
Lithium acetate 1 mol/l in aqueous solution
Catalog # 82023-112
Supplier:  G-Biosciences
Supplier Number:  R039
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 ml
  • Cat. No.:
    82023-112
  • Supplier no.:
    R039

Specifications

About this item

 :
 :65.99 g/mol
Storage Temperature:Ambient
EINECS:208-914-3
CAS Number:546-89-4
Formula:H₃CCOOLi
MDL Number:MFCD00013057