ContactUsLinkComponent
HardyCHROM™ MRSA (Methicillin-Resistant Staphylococcus aureus), Hardy Diagnostics
HardyCHROM™ MRSA (Methicillin-Resistant Staphylococcus aureus), Hardy Diagnostics
Catalog # 76359-396
Supplier:  Hardy Diagnostics
CAS Number:  
undefined
HardyCHROM™ MRSA (Methicillin-Resistant Staphylococcus aureus), Hardy Diagnostics
Catalog # 76359-396
Supplier:  Hardy Diagnostics
Supplier Number:  GA307
CAS Number:  
Specifications
  • Description:
    HardyCHROM™ MRSA (methicillin-resistant Staphylococcus aureus)
  • Size:
    10/pack
  • Cat. No.:
    76359-396
  • Plate size:
    15×100 mm
Specifications
About this item
HardyCHROM™ MRSA is a selective and differential culture medium that facilitates the isolation and identification of methicillin resistant Staphylococcus aureus (MRSA) to aid in the prevention and control of MRSA infections in health care settings. The test is performed on anterior nares swabs from patients and healthcare workers to screen for MRSA colonization.
  • Distinct color change read-out
  • Bright color development
  • Compatible with automation
  • Bright color development in 24 hours
This new chromogenic medium simplifies the identification of MRSA infections. MRSA strains grown in the presence of chromogenic substrates produce deep pink to magenta colonies. Color development is bright, distinct and easy to read.
Caution: This media is to be stored and incubated in the dark. If you have a light in your refrigerator, incubator, or you leave the media exposed to light on the bench top, Hardy Diagnostics offers free Blok-Boxes upon request. Blok-Box is a reusable cardboard box that holds one sleeve of ten petri plates.