ContactUsLinkComponent
PPLO Selective Agar, for Mycoplasma pneumoniae, Hardy Diagnostics
PPLO Selective Agar, for Mycoplasma pneumoniae, Hardy Diagnostics
Catalog # 76479-596
Supplier:  Hardy Diagnostics
CAS Number:  
undefined
PPLO Selective Agar, for Mycoplasma pneumoniae, Hardy Diagnostics
Catalog # 76479-596
Supplier:  Hardy Diagnostics
Supplier Number:  G105
CAS Number:  
Specifications
  • Description:
    Prepared media plate for Mycoplasma pneumoniae
  • Size:
    10/pack
  • Cat. No.:
    76479-596
  • Tube size:
    15×100 mm
Specifications
About this item
For the selective isolation of Mycoplasmataceae (pleural pneumonia-like organisms), especially Mycoplasma pneumoniae.