ContactUsLinkComponent
Orum International Power Supply Charging Cord for TRIO.BAS™ Instruments, Hardy Diagnostics
Orum International Power Supply Charging Cord for TRIO.BAS™ Instruments, Hardy Diagnostics
Catalog # 76446-892
Supplier:  Hardy Diagnostics
undefined
Orum International Power Supply Charging Cord for TRIO.BAS™ Instruments, Hardy Diagnostics
Catalog # 76446-892
Supplier:  Hardy Diagnostics
Supplier Number:  BAS621
Specifications
  • Voltage:
    220/180 V
  • Description:
    Charging cord
  • Cat. No.:
    76446-892
Specifications
About this item
Wall charger power supply (charging cord) for TRIO.BAS™ MINI, and TRIO.BAS™ instruments using cables rather than the induction battery charger system.
  • 220/180 V power supply