ContactUsLinkComponent
HOBO® Water Level 13' Data Logger
HOBO® Water Level 13' Data Logger
Catalog # 470323-414
undefined
HOBO® Water Level 13' Data Logger
Catalog # 470323-414
Supplier Number:  U20L-04
Specifications
  • Description:
    HOBO® water level 13' data logger
  • Cat. No.:
    470323-414
Specifications
About this item
The HOBO® U20L-04 is a low-cost, research-grade water level data logger for continuously measuring water level and temperature in a wide range of underwater environments.
  • Self-contained non-vented design enables easy deployment
  • Ideal for use in both fresh and saltwater environments, including wells, streams, lakes, wetlands, and tidal areas
  • Depths up to 13 feet
  • Durable ceramic pressure sensor withstands freezing
  • HOBOware® Pro software provides easy conversion to accurate water level reading, fully compensated for barometric pressure, temperature, and water density
It features 0.1% measurement accuracy, a polypropylene housing for use in both fresh and salt water, and a non-vented design for convenient and hassle-free deployment.
Ordering information: This logger requires HOBOware® Pro software and either a U-DTW-1 Waterproof Shuttle or the Base-U-4 Optic USB Base Station for configuration and data offload. For more information on required HOBOware software (not included), contact your Avantor representative.
Sotware
Product Information