Human Recombinant CD39 (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris and NaCl, pH8.0. Normally Trehalose is added as protectant before lyophilization.
- Gene ID:P49961-1;953;NP_001767.3
- Protein Synonyms:CD39,ENTPD1,NTPDase 1,Entpd1,Ecto-ATPDase 1,Ecto-ATPase 1
- Protein/Peptide Name:CD39
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:52.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris and NaCl, pH8.0. Normally Trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together