Order Entry
United States
ContactUsLinkComponent
Human Recombinant LAG-3 (from HEK293), PE Labeled Protein
Human Recombinant LAG-3 (from HEK293), PE Labeled Protein
Supplier:  ACROBIOSYSTEMS
Certificates
Human Recombinant LAG-3 (from HEK293), PE Labeled Protein
Supplier:  ACROBIOSYSTEMS

Specifications

  • Species:
    Human
  • Storage Conditions:
    –20 °C, aviod light
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS with Arginine, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P18627-1;3902;NP_002277.4
  • Protein Synonyms:
    LAG3,CD223,FDC
  • Protein/Peptide Name:
    LAG-3
  • Molecular Weight:
    49.9 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS with Arginine, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together