Human Recombinant CA125 (from HEK293), Precision Avi
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4.
- Gene ID:Q8WXI7-1;94025;NP_078966.2
- Protein Synonyms:CA125,CA-125,CA125MUC-16,FLJ14303,MUC16,mucin 16,mucin-16
- Protein/Peptide Name:CA125
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:100.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4.
- Shipping Temperature:RT
Specifications
Frequently Bought Together