Cynomolgus Recombinant CLEC12A (from HEK293), Unconjugated
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Cynomolgus monkey
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally Trehalose is added as protectant before lyophilization.
- Gene ID:A0A2K5WXQ6-1
- Protein Synonyms:CLEC12A,MICL,CLL-1,CLL1,DCAL2,DCAL-2,CD371
- Protein/Peptide Name:CLEC12A
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:25.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally Trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together