SARS-CoV-2 Recombinant Nucleocapsid protein (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:SARS-CoV-2
- Storage Conditions:–70℃
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Supplied as 0.2 μm filtered solution in 10 mM PB, 150 mM NaCl, pH7.4.
- Gene ID:YP_009724397.2
- Protein Synonyms:Nucleocapsid protein,NP,Protein N
- Protein/Peptide Name:Nucleocapsid protein
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:47.3 kDa
- Endotoxin Level:Low
- Formulation:Supplied as 0.2 μm filtered solution in 10 mM PB, 150 mM NaCl, pH7.4.
- Shipping Temperature:Dry Ice
Specifications
Frequently Bought Together