Human Recombinant IL-12B & IL-12A (from HEK293), Precision Avi
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:P29460-1 (IL12B) & P29459-1 (IL12A)
- Protein Synonyms:IL12,p70,Interleukin-12
- Protein/Peptide Name:IL-12B & IL-12A
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:38.3 kDa | 23.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together