Order Entry
United States
ContactUsLinkComponent
Cynomolgus Recombinant IL-1 Rrp2 (from HEK293), Unconjugated
Cynomolgus Recombinant IL-1 Rrp2 (from HEK293), Unconjugated
Supplier:  ACRO Biosystems
Certificates
Cynomolgus Recombinant IL-1 Rrp2 (from HEK293), Unconjugated
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Cynomolgus monkey
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    EHH61702.1
  • Protein Synonyms:
    IL-1 R6,IL-1 Rrp2,IL1RL2,IL-1RL2,IL1Rrp2,IL-1Rrp2,IL1RRP2,IL-36R
  • Protein/Peptide Name:
    IL-1 Rrp2
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    37.9 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together