Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant Syndecan-1 (from HEK293), FITC Labeled Protein
 
Human Recombinant Syndecan-1 (from HEK293), FITC Labeled Protein

 

  • Species:
    Human
  • Storage Conditions:
    –20 °C, aviod light
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; 2e5 of anti-Syndecan-1 CAR-293 cells were stained with 100 μL of 3 μg/mL of FITC-Labeled Human Syndecan-1, His Tag (Cat. No. SD1-HF228) and negative control protein respectively, FITC signals was used to evaluate the binding activity (QC tested).
  • Gene ID:
    P18827-1;6382;NP_001006947.1(L136Q)
  • Protein Synonyms:
    SDC1,Syndecan-1,CD138,SYND1,SDC
  • Protein/Peptide Name:
    Syndecan-1
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    25.8 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

 

Frequently Bought Together