Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD5 (from HEK293), FITC Labeled Protein
Human Recombinant CD5 (from HEK293), FITC Labeled Protein
Supplier:  ACRO Biosystems
Certificates
Human Recombinant CD5 (from HEK293), FITC Labeled Protein
Supplier:  ACRO Biosystems

Specifications

  • Species:
    Human
  • Storage Conditions:
    –20 °C, aviod light
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P06127-1
  • Protein Synonyms:
    CD5,LEU1
  • Protein/Peptide Name:
    CD5
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    40.5 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together