Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant CLEC12A (from HEK293), Precision Avi
 
Human Recombinant CLEC12A (from HEK293), Precision Avi

 

  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; 2e5 of anti-CLEC12A CAR-293 cells were stained with 100 μL of 10 μg/mL of Biotinylated Human CLEC12A, His,Avitag (Cat. No. CLA-H82E6) and negative control protein respectively, washed and then followed by PE-SA and analyzed with FACS (Routinely tested).
  • Gene ID:
    Q5QGZ9-2;160364;NP_612210.4
  • Protein Synonyms:
    CLEC12A,MICL,CLL-1,CLL1,DCAL2,DCAL-2,CD371
  • Protein/Peptide Name:
    CLEC12A
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    27.3 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

 

Frequently Bought Together