Cynomolgus Recombinant CD3E and CD3G (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Cynomolgus monkey
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Loaded Cynomolgus CD3E&CD3G Heterodimer Protein, Fc Tag&Fc Tag (Cat. No. CDG-C5257) on AHC Biosensor, can bind Human CD3×BCMA scFV with an affinity constant of 1.02 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q95LI5-1 (CD3E) & Q95LI7-1 (CD3G);102133065 | 102134381;NP_001270544.1 (CD3E) & NP_001270839.1 (CD3G)
- Protein Synonyms:CD3 epsilon & CD3 gamma,CD3E & CD3G
- Protein/Peptide Name:CD3E & CD3G
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:40.6 kDa (CD3E) & 40.4 kDa (CD3G)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together