Human Recombinant IL-17 RA & IL-17 RC (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.; Loaded Human IL17RA & IL17RC Protein, Fc Tag&Fc Tag (Cat. No. ILC-H5257) on Protein A Biosensor, can bind Human IL17A, Tag Free (Cat. No. ILA-H5118) with an affinity constant of 0.13 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q96F46-1 (IL-17RA) & NP_703190.1 (IL-17RC)
- Protein Synonyms:IL-17 RA & IL-17 RC,IL-17RA & IL-17RC,CD217 & IL-17 RC
- Protein/Peptide Name:IL-17 RA & IL-17 RC
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:60.0 kDa (IL-17 RA) & 75.7 kDa (IL-17 RC)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together