Order Entry
United States
ContactUsLinkComponent
SARS-CoV-2 Recombinant S protein RBD (from HEK293), Precision Avi
SARS-CoV-2 Recombinant S protein RBD (from HEK293), Precision Avi
Supplier:  ACROBIOSYSTEMS
Certificates
SARS-CoV-2 Recombinant S protein RBD (from HEK293), Precision Avi
Supplier:  ACROBIOSYSTEMS

Specifications

  • Species:
    SARS-CoV-2
  • Storage Conditions:
    –70℃
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Supplied as 0.2 μm filtered solution in 10 mM PB, pH7.4.
  • Gene ID:
    QHD43416.1
  • Protein Synonyms:
    Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD
  • Protein/Peptide Name:
    S protein RBD
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    28.2 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Supplied as 0.2 μm filtered solution in 10 mM PB, pH7.4.
  • Shipping Temperature:
    Dry Ice

Specifications

Frequently Bought Together