Mouse Recombinant CD3E & CD3D (from HEK293), Precision Avi
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:P22646-1 (CD3E) & P04235-1 (CD3D)
- Protein Synonyms:CD3E & CD3D,CD3 delta & CD3 epsilon,CD3 delta , epsilon
- Protein/Peptide Name:CD3E & CD3D
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:17.3 kDa (CD3E) & 13.3 kDa (CD3D)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together