Human Recombinant NOTCH2 (from HEK293), Unconjugated
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Loaded Human NOTCH2, Fc Tag (Cat. No. NO2-H5255) on Protein A Biosensor, can bind Human DLL4, His Tag (Cat. No. DL4-H5227) with an affinity constant of 77.6 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q04721-1;4853;NP_077719.2
- Protein Synonyms:NOTCH2,hN2,N2ECD
- Protein/Peptide Name:NOTCH2
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:80.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together