Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant IL-6 R alpha (from HEK293), Precision Avi
Mouse Recombinant IL-6 R alpha (from HEK293), Precision Avi
Supplier:  ACRO Biosystems
Certificates
Mouse Recombinant IL-6 R alpha (from HEK293), Precision Avi
Supplier:  ACRO Biosystems

Specifications

  • Species:
    Mouse
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P22272-1;16194;NP_034689.2
  • Protein Synonyms:
    IL6R,CD126,IL-6R-1,IL-6RA,IL6RA,IL-6R-alpha,IL6RQ,gp80
  • Protein/Peptide Name:
    IL-6 R alpha
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    41.0 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together