HCoV-229E Recombinant Nucleocapsid protein (from E.coli), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:HCoV-229E
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 40 mM PB, 500 mM NaCl, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:P15130-1
- Protein Synonyms:Nucleocapsid protein,NP,Protein N
- Protein/Peptide Name:Nucleocapsid protein
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:44.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 40 mM PB, 500 mM NaCl, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together