Human Recombinant EpCAM (from HEK293), FITC Labeled Protein
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Species:Human
- Storage Conditions:–20 °C, aviod light
- Biological Activity:Please contact us for detailed information.
- Gene ID:P16422-1(M115T);4072;AAH14785.1
- Protein Synonyms:EPCAM,TACSTD1,TROP1,CD326,DIAR5,EGP2,EGP314,EGP40,ESA,GA733-2,HNPCC8,HNPCC-8,KS1,4,KSA,M4S1,MIC18,MK1
- Protein/Peptide Name:EpCAM
- Molecular Weight:28.2 kDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:RT
Specifications
Frequently Bought Together