Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant FAP (from HEK293), Precision Avi
Mouse Recombinant FAP (from HEK293), Precision Avi
Supplier:  ACROBIOSYSTEMS
Certificates
Mouse Recombinant FAP (from HEK293), Precision Avi
Supplier:  ACROBIOSYSTEMS

Specifications

  • Species:
    Mouse
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P97321
  • Protein Synonyms:
    FAP,FAPalpha,SIMP,Seprase,APCE
  • Protein/Peptide Name:
    FAP
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    88.8 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together