Human Recombinant CD3E & CD3D (from HEK293), Precision Avi
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Loaded Biotinylated Human CD3E&CD3D Heterodimer Protein, Fc,His,Avitag&Fc,Flag,Avitag (Cat. No. CDD-H82F6) on SA Biosensor, can bind Bispecific CD3×BCMA T cell-engaging Antibody with an affinity constant of 1.4 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Biotinylated Human CD3E&CD3D Heterodimer Protein, Fc,His,Avitag&Fc,Flag,Avitag (Cat. No. CDD-H82F6) on SA Biosensor, can bind UCHT1 with an affinity constant of 21.3 pM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Biotinylated Human CD3E&CD3D Heterodimer Protein, Fc,His,Avitag&Fc,Flag,Avitag (Cat. No. CDD-H82F6) on SA Biosensor, can bind Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a (Clone: OKT3), Ultra-low endotoxin (Cat. No. CDE-M120a) with an affinity constant of 1.5 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P07766-1 (CD3E) & P04234-1 (CD3D);NP_000724.1 (CD3E) & NP_000723.1 (CD3D)
- Protein Synonyms:CD3E & CD3D,CD3 delta & CD3 epsilon,CD3 delta , epsilon
- Protein/Peptide Name:CD3E & CD3D
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:45.1kDa (CD3E) & 42.6 kDa (CD3D)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Frequently Bought Together