Order Entry
United States
ContactUsLinkComponent
Human Recombinant CCR5 (from HEK293), Unconjugated
Human Recombinant CCR5 (from HEK293), Unconjugated
Supplier:  ACROBIOSYSTEMS
Certificates
Human Recombinant CCR5 (from HEK293), Unconjugated
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Storage Conditions:
    –70℃
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Supplied as 0.2 μm filtered solution in 50 mM HEPES, 150 mM NaCl, DDM, CHS, pH7.5 with glycerol as protectant.; Human CCR5, Flag,His Tag (Cat. No. CC5-H52D1) captured on CM5 chip via anti-His antibody can bind Anti-CCR5 antibody (Human IgG1) with an affinity constant of 0.0737 nM as determined in a SPR assay (in presence of DDM and CHS) (Biacore T200) (Routinely tested).; Purified NA/LE Mouse Anti-Human CD195 (Clone: 2D7) (Mouse IgG2a) captured on CM5 chip via anti-mouse antibodies surface can bind Human CCR5, Flag,His Tag (Cat. No. CC5-H52D1) with an affinity constant of 7.51 nM as determined in a SPR assay (in presence of DDM and CHS) (Biacore T200) (Routinely tested).
  • Gene ID:
    P51681-1 (C58Y, G163N, A233D, K303E)
  • Protein Synonyms:
    CCR5,CMKBR5,CD195,CC-CKR-5,CHEMR13
  • Protein/Peptide Name:
    CCR5
  • Purity:
    >85% as determined by SDS-PAGE.
  • Molecular Weight:
    46.0 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Supplied as 0.2 μm filtered solution in 50 mM HEPES, 150 mM NaCl, DDM, CHS, pH7.5 with glycerol as protectant.
  • Shipping Temperature:
    Dry Ice

Specifications

Frequently Bought Together