Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant TGF-beta 1 (from HEK293), Unconjugated
Mouse Recombinant TGF-beta 1 (from HEK293), Unconjugated
Supplier:  ACROBIOSYSTEMS
Certificates
Mouse Recombinant TGF-beta 1 (from HEK293), Unconjugated
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Mouse
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in 50 mM HAC, pH3.0. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P04202-1;21803;NP_035707.1
  • Protein Synonyms:
    CEDLAP,TGF-beta 1,TGFB1,DPD1,TGF-beta-1,TGFB
  • Protein/Peptide Name:
    TGF-beta 1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    12.8 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 50 mM HAC, pH3.0. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together