Human Recombinant B7-H6 (from HEK293), Precision Avi
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
- Gene ID:Q68D85-1;NP 001189368.1
- Protein Synonyms:B7-H6,NCR3LG1,B7 Homolog 6
- Protein/Peptide Name:B7-H6
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:54.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together