Human Recombinant ASGR1 (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Loaded Human ASGR1, Fc Tag (Cat. No. AS1-H5253) on Protein A Biosensor, can bind SARS-CoV-2 S1 protein NTD, His Tag (Cat. No. S1D-C52H6) with an affinity constant of 401 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Human ASGR1, Fc Tag (Cat. No. AS1-H5253) on Protein A Biosensor, can bind SARS-CoV-2 S1 protein with an affinity constant of 4.09 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Human ASGR1, Fc Tag (Cat. No. AS1-H5253) on Protein A Biosensor, can bind SARS-CoV-2 S protein RBD, His Tag (Cat. No. SPD-C52H3) with an affinity constant of 686 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P07306-1;432;NP_001662.1
- Protein Synonyms:ASGP-R 1, ASGPR 1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1
- Protein/Peptide Name:ASGR1
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:52.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together