Order Entry
United States
ContactUsLinkComponent
Cynomolgus Recombinant Fc gamma RIIB / CD32b (from HEK293), Unconjugated
Cynomolgus Recombinant Fc gamma RIIB / CD32b (from HEK293), Unconjugated
Supplier:  ACROBIOSYSTEMS
Certificates
Cynomolgus Recombinant Fc gamma RIIB / CD32b (from HEK293), Unconjugated
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Cynomolgus monkey
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Cynomolgus CD32b, His Tag (Cat. No. CDB-C5224) captured on CM5 chip via anti-His antibody can Herceptin® with an affinity constant of 11.9 μM as determined in a SPR assay (Biacore T200) (QC tested).
  • Gene ID:
    Q8SPW3-1;102139530;NP_001271060.1
  • Protein Synonyms:
    Fc gamma RIIB,CD32b
  • Protein/Peptide Name:
    Fc gamma RIIB / CD32b
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    21.8 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together