Mouse Recombinant CD3E & CD3D (from HEK293), Unconjugated
- Conjugation:Unconjugated
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100mM Glycine, 25mM Arginine, 150mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Gene ID:P22646-1 (CD3E) & P04235-1 (CD3D);12501 | 12500;NP_031674.1 (CD3E) & NP_038515.3 (CD3D)
- Protein Synonyms:CD3E & CD3D,CD3 delta & CD3 epsilon,CD3 delta , epsilon
- Protein/Peptide Name:CD3E & CD3D
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:41.5 kDa (CD3E) & 40.7 kDa (CD3D)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100mM Glycine, 25mM Arginine, 150mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Frequently Bought Together