Human Recombinant EMMPRIN (from HEK293), FITC Labeled Protein
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Species:Human
- Storage Conditions:–20 °C, aviod light
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:P35613-2;682;NP_940991.1
- Protein Synonyms:Basigin,BSG,5F7,CD147,EMMPRIN,M6,OK,TCSF
- Protein/Peptide Name:EMMPRIN
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:20.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together