Cynomolgus Recombinant Mucin-1 (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Cynomolgus monkey
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 25 mM Arginine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Gene ID:102123068;XP_005541632.1
- Protein Synonyms:Mucin 1,MUC1,CD227,EMA,H23AG,KL-6,MAM6,MUC-1,SEC,MUC-1,X,MUC1,ZD,PEM,PEMT,PUM,CA 15-3,Episialin
- Protein/Peptide Name:Mucin-1
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:7.2 kDa and 32.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 25 mM Arginine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together