Rhesus macaque / Cynomolgus Recombinant TROP-2 (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:102127436;XP_001114599.1
- Protein Synonyms:TACSTD2,GA733-1,M1S1,TROP2
- Protein/Peptide Name:TROP-2
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:29.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together