Human Recombinant CD5 (from HEK293), Precision Avi
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:P06127-1;921;NP_055022.2
- Protein Synonyms:CD5,LEU1
- Protein/Peptide Name:CD5
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:42.2 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together