Human Recombinant IL-13 R alpha 2 (from HEK293), Precision Avi
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:Q14627-1;3598;NP_000631.1
- Protein Synonyms:IL13RA2,CD213A2,CT19,IL-13R,IL13BP
- Protein/Peptide Name:IL-13 R alpha 2
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:40.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together