Human Recombinant LY75 (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:O60449-1;4065;NP_002340.2
- Protein Synonyms:LY75,CD205,DEC-205,CLEC13B,Lymphocyte antigen 75,gp200-MR6
- Protein/Peptide Name:LY75
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:190.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together