Human Recombinant MARCO (from HEK293), Precision Avi
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Species:Human
- Storage Conditions:–20 °C
- Biological Activity:Please contact us for detailed information.
- Gene ID:Q9UEW3-1
- Protein Synonyms:MARCO, SCARA2, Macrophage receptor with collagenous structure, Scavenger receptor class A member 2
- Protein/Peptide Name:MARCO
- Molecular Weight:46.8 KDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:RT
Specifications
Frequently Bought Together