Order Entry
United States
ContactUsLinkComponent
Streptococcus pyogenes Recombinant IdeS (from E.coli), Unconjugated
Streptococcus pyogenes Recombinant IdeS (from E.coli), Unconjugated
Supplier:  ACROBIOSYSTEMS
Certificates
Streptococcus pyogenes Recombinant IdeS (from E.coli), Unconjugated
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Streptococcus pyogenes
  • Storage Conditions:
    –70℃
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Supplied as 0.2 μm filtered solution in PBS, pH7.4 with glycerol as protectant.
  • Gene ID:
    Q9F1R7-1;WP_010922160.1
  • Protein Synonyms:
    IgG-degrading protease,Sib38,IdeS
  • Protein/Peptide Name:
    IdeS
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    36.8 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Supplied as 0.2 μm filtered solution in PBS, pH7.4 with glycerol as protectant.
  • Shipping Temperature:
    Dry Ice

Specifications

Frequently Bought Together