Order Entry
United States
ContactUsLinkComponent
Human Recombinant Her2 (from HEK293), Unconjugated
Human Recombinant Her2 (from HEK293), Unconjugated
Supplier:  ACRO Biosystems
Certificates
Human Recombinant Her2 (from HEK293), Unconjugated
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P04626-1;2064;NP_004439.2
  • Protein Synonyms:
    ERBB2,CD340,HER-2,neu,HER2,MLN19,NEU,NGL,TKR1
  • Protein/Peptide Name:
    Her2
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    55.6 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together