Human Recombinant IL-34 (from HEK293), Unconjugated
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:Q6ZMJ4-1;146433;NP_689669.2
- Protein Synonyms:IL34,C16orf77,IL-34,Interleukin-34,MGC34647
- Protein/Peptide Name:IL-34
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:27.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together